Methyl 3-Bromo-5-(hydroxymethyl)benzoate
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-bromo-5-(hydroxymethyl)benzoate
OR400939-1G
OR400939-250MG
APOSOR400939-1GEA
207
EUR
APOSOR400939-1G
APOSOR400939-250MG
Methyl 3-Bromo-5-(hydroxymethyl)benzoate
Methyl 3-Bromo-5-(hydroxymethyl)benzoate
Formel:
C₉H₉BrO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11040228 CAS-Nummer: 307353-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...