2-Formylphenyl acetate, tech

Lieferant: Apollo Scientific
Achtung

OR400800-10G OR400800-1G OR400800-5G
APOSOR400800-10GEA 662 EUR
APOSOR400800-10G APOSOR400800-1G APOSOR400800-5G
2-Formylphenyl acetate, tech
2-Formylphenyl acetate
Formel: C₉H₈O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00553742
CAS-Nummer: 5663-67-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR