4-(Dimethylamino)benzyl bromide hydrobromide, tech
Lieferant: Apollo Scientific
|
Gefahr
|
OR400627-1G
OR400627-250MG
APOSOR400627-1GEA
522
EUR
APOSOR400627-1G
APOSOR400627-250MG
4-(Dimethylamino)benzyl bromide hydrobromide, tech
4-(Dimethylamino)benzyl bromide hydrobromide
Formel:
C₉H₁₂BrN·BrH Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD13195837 CAS-Nummer: 1116572-69-0 UN-Nummer: 2928 ADR: 6.1+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...