4-Bromo-N,3-dimethyl-N-methoxybenzamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-N-methoxy-N,3-dimethylbenzamide

OR400581-1G OR400581-5G
APOSOR400581-1GEA 464 EUR
APOSOR400581-1G APOSOR400581-5G
4-Bromo-N,3-dimethyl-N-methoxybenzamide
4-Bromo-N,3-dimethyl-N-methoxybenzamide
Formel: C₁₀H₁₂BrNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD26802226
CAS-Nummer: 170230-01-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR