2-Amino-5-bromo-4-methoxybenzoic acid

Lieferant: Apollo Scientific
Achtung

OR400461-250MG OR400461-1G OR400461-5G
APOSOR400461-250MGEA 58 EUR
APOSOR400461-250MG APOSOR400461-1G APOSOR400461-5G
2-Amino-5-bromo-4-methoxybenzoic acid
2-Amino-5-bromo-4-methoxybenzoic acid
Formel: C₈H₈BrNO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD21868786
CAS-Nummer: 169045-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR