3'-Bromo-4'-iodoacetophenone

Lieferant: Apollo Scientific
Achtung

OR400426-1G OR400426-250MG OR400426-5G
APOSOR400426-1GEA 29 EUR
APOSOR400426-1G APOSOR400426-250MG APOSOR400426-5G
3'-Bromo-4'-iodoacetophenone
3'-Bromo-4'-iodoacetophenone
Formel: C₈H₆BrIO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18391610
CAS-Nummer: 945907-32-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR