3-Piperidin-4-yl-1H-quinolin-2-one ≥98%
Lieferant: Apollo Scientific
OR370039-100MG
OR370039-1G
OR370039-250MG
APOSOR370039-100MGEA
277
EUR
APOSOR370039-100MG
APOSOR370039-1G
APOSOR370039-250MG
3-Piperidin-4-yl-1H-quinolin-2-one ≥98%
3-Piperidin-4-yl-1H-quinolin-2-one
Formel:
C₁₄H₁₆N₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD15527014 CAS-Nummer: 205058-78-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...