1-(4-Ethynyl-2-methoxyphenyl)-4-methyl-1H-imidazole ≥98%
Lieferant: Apollo Scientific
OR370024-100MG
OR370024-500MG
APOSOR370024-100MGEA
598
EUR
APOSOR370024-100MG
APOSOR370024-500MG
1-(4-Ethynyl-2-methoxyphenyl)-4-methyl-1H-imidazole ≥98%
1-(4-Ethynyl-2-methoxyphenyl)-4-methyl-1H-imidazole
Formel:
C₁₃H₁₂N₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12545876 CAS-Nummer: 1093980-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...