4-(tert-Butoxycarbonyl-n-methylamino)-phenylboronic acid 98%
Lieferant: Apollo Scientific
Synonyme:
(4-((tert-Butoxycarbonyl)(methyl)amino)phenyl)boronic acid
OR361605-1G
OR361605-5G
APOSOR361605-1GEA
116
EUR
APOSOR361605-1G
APOSOR361605-5G
4-(tert-Butoxycarbonyl-n-methylamino)-phenylboronic acid 98%
4-(tert-Butoxycarbonyl-n-methylamino)-phenylboronic acid
Formel:
C₁₂H₁₈BNO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03095105 CAS-Nummer: 945756-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...