Methyl 4-(hydroxymethyl)bicyclo[2.2.2]octane-1-carboxylate
Lieferant: Apollo Scientific
OR350282-1G
OR350282-250MG
APOSOR350282-1GEA
242
EUR
APOSOR350282-1G
APOSOR350282-250MG
Methyl 4-(hydroxymethyl)bicyclo[2.2.2]octane-1-carboxylate
Methyl 4-(hydroxymethyl)bicyclo[2.2.2]octane-1-carboxylate
Formel:
C₁₁H₁₈O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD20275559 CAS-Nummer: 94994-15-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...