3-Chloro-4-(methoxycarbonyl)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR350093-250MG
OR350093-1G
APOSOR350093-250MGEA
59.6
EUR
APOSOR350093-250MG
APOSOR350093-1G
3-Chloro-4-(methoxycarbonyl)benzoic acid 97%
3-Chloro-4-(methoxycarbonyl)benzoic acid
Formel:
C₉H₇ClO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00100478 CAS-Nummer: 55737-77-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...