(4-Amino-phenyl)-methyl-carbamic acid tert-butyl ester

Lieferant: Apollo Scientific

OR346359-1G OR346359-5G
APOSOR346359-1GEA 0 EUR
APOSOR346359-1G APOSOR346359-5G
(4-Amino-phenyl)-methyl-carbamic acid tert-butyl ester
(4-Amino-phenyl)-methyl-carbamic acid tert-butyl ester
Formel: C₁₂H₁₈N₂O₂
MDL Number: MFCD10693199
CAS-Nummer: 1092522-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR