5-sulphamoyl-furan-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-sulphamoylfuran-3-carboxylate, Ethyl 5-sulphamoyl-3-furoate

OR346356-1G OR346356-250MG
APOSOR346356-1GEA 212 EUR
APOSOR346356-1G APOSOR346356-250MG
5-sulphamoyl-furan-3-carboxylic acid ethyl ester
5-sulphamoyl-furan-3-carboxylic acid ethyl ester
Formel: C₇H₉NO₅S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD24386139
CAS-Nummer: 256373-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR