4-(3-Methyl-isothiazol-5-yl)-benzoic acid methyl ester

Lieferant: Apollo Scientific

OR346315-1G OR346315-500MG
APOSOR346315-1GEA 615 EUR
APOSOR346315-1G APOSOR346315-500MG
4-(3-Methyl-isothiazol-5-yl)-benzoic acid methyl ester
Formel: C₁₂H₁₁NO₂S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD22573493
CAS-Nummer: 39101-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR