3-Bromo-4-oxo-piperidine-1-carboxylic acid benzyl ester
Lieferant: Apollo Scientific
Synonyme:
Benzyl 3-bromo-4-oxopiperidine-1-carboxylate
OR346180-1G
OR346180-5G
APOSOR346180-1GEA
0
EUR
APOSOR346180-1G
APOSOR346180-5G
3-Bromo-4-oxo-piperidine-1-carboxylic acid benzyl ester
3-Bromo-4-oxo-piperidine-1-carboxylic acid benzyl ester
Formel:
C₁₃H₁₄BrNO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09953304 CAS-Nummer: 174184-13-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...