2-Amino-4-methyl-5-pyridinecarboxylic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Amino-4-methylnicotinic acid
OR345282-1G
OR345282-250MG
OR345282-5G
APOSOR345282-1GEA
79.9
EUR
APOSOR345282-1G
APOSOR345282-250MG
APOSOR345282-5G
2-Amino-4-methyl-5-pyridinecarboxylic acid 95+%
2-Amino-4-methyl-5-pyridinecarboxylic acid
Formel:
C₇H₈N₂O₂ Schmelzpunkt: 270…272 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09835242 CAS-Nummer: 179555-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...