[5-(Fur-2-yl)isoxazol-3-yl]methylamine hydrochloride

Lieferant: Apollo Scientific
Gefahr

OR33648-1G OR33648-250MG OR33648-50MG OR33648-100MG OR33648-500MG
APOSOR33648-1GEA 0 EUR
APOSOR33648-1G APOSOR33648-250MG APOSOR33648-50MG APOSOR33648-100MG APOSOR33648-500MG
[5-(Fur-2-yl)isoxazol-3-yl]methylamine hydrochloride
[5-(Fur-2-yl)isoxazol-3-yl]methylamine hydrochloride
Formel: C₈H₈N₂O₂·ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16660999
CAS-Nummer: 1255717-94-2
UN-Nummer: 3259
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR