3-(Dimethylamino)-1-(4-methylphenyl)propan-1-one hydrochloride 95%

Lieferant: Apollo Scientific

OR33192-1G OR33192-5G
APOSOR33192-1GEA 224 EUR
APOSOR33192-1G APOSOR33192-5G
3-(Dimethylamino)-1-(4-methylphenyl)propan-1-one hydrochloride 95%
3-(Dimethylamino)-1-(4-methylphenyl)propan-1-one hydrochloride
Formel: C₁₂H₁₇NO·ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00156711
CAS-Nummer: 5250-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR