4-Chloro-2-methyl-1H-1,3-benzodiazole 95%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-2-methyl-1H-benzo[d]imidazole
OR33139-1G
OR33139-5G
APOSOR33139-1GEA
339
EUR
APOSOR33139-1G
APOSOR33139-5G
4-Chloro-2-methyl-1H-1,3-benzodiazole 95%
4-Chloro-2-methyl-1H-1,3-benzodiazole
Formel:
C₈H₇ClN₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02572038 CAS-Nummer: 5599-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...