7-Methyl-1H-indazole-5-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR322740-100MG
OR322740-1G
OR322740-250MG
APOSOR322740-100MGEA
101
EUR
APOSOR322740-100MG
APOSOR322740-1G
APOSOR322740-250MG
7-Methyl-1H-indazole-5-carboxylic acid
7-Methyl-1H-indazole-5-carboxylic acid
Formel:
C₉H₈N₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD14585353 CAS-Nummer: 1031417-41-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...