3-(1H-Pyrazol-4-yl)piperidine dihydrochloride

Lieferant: Apollo Scientific

OR321519-1G OR321519-250MG
APOSOR321519-1GEA 980 EUR
APOSOR321519-1G APOSOR321519-250MG
3-(1H-Pyrazol-4-yl)piperidine dihydrochloride
3-(1H-Pyrazol-4-yl)piperidine dihydrochloride
Formel: C₈H₁₃N₃·2ClH
Lagerungstemperatur: Kühlschrank
CAS-Nummer: 2095410-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR