2-(Piperidin-2-yl)pyridine dihydrochloride 95%

Lieferant: Apollo Scientific

OR321385-100MG OR321385-250MG OR321385-1G
APOSOR321385-100MGEA 43.5 EUR
APOSOR321385-100MG APOSOR321385-250MG APOSOR321385-1G
2-(Piperidin-2-yl)pyridine dihydrochloride 95%
2-(Piperidin-2-yl)pyridine dihydrochloride
Formel: C₁₀H₁₄N₂·2ClH
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD08461227
CAS-Nummer: 143924-44-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR