6-Bromo-1H-indazole-3-carbonitrile 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR321320-1G
OR321320-250MG
APOSOR321320-1GEA
150
EUR
APOSOR321320-1G
APOSOR321320-250MG
6-Bromo-1H-indazole-3-carbonitrile 97%
6-Bromo-1H-indazole-3-carbonitrile
Formel:
C₈H₄BrN₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06659785 CAS-Nummer: 885278-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...