Adamantane-1-carboxaldehyde
Lieferant: Apollo Scientific
Synonyme:
1-Adamantanecarboxaldehyde
OR315781-1G
OR315781-250MG
APOSOR315781-1GEA
139
EUR
APOSOR315781-1G
APOSOR315781-250MG
Adamantane-1-carboxaldehyde
Adamantane-1-carboxaldehyde
Formel:
C₁₁H₁₆O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12031698 CAS-Nummer: 2094-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...