2-Methyl-3-hydroxy-6-iodopyridine 95%

Lieferant: Apollo Scientific

Synonyme: 6-Iodo-2-methylpyridin-3-ol

OR314076-1G OR314076-250MG OR314076-5G
APOSOR314076-1GEA 200 EUR
APOSOR314076-1G APOSOR314076-250MG APOSOR314076-5G
2-Methyl-3-hydroxy-6-iodopyridine 95%
2-Methyl-3-hydroxy-6-iodopyridine
Formel: C₆H₆INO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11044376
CAS-Nummer: 848952-39-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR