(2R,3R,5R,6R,7R,8R)-Cubane-1-carboxylic acid 95%

Lieferant: Apollo Scientific

OR312529-1G OR312529-250MG
APOSOR312529-1GEA 2670 EUR
APOSOR312529-1G APOSOR312529-250MG
(2R,3R,5R,6R,7R,8R)-Cubane-1-carboxylic acid 95%
(2R,3R,5R,6R,7R,8R)-Cubane-1-carboxylic acid
Formel: C₉H₈O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD29060095
CAS-Nummer: 53578-15-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR