(4-Benzylphenyl)methanol

Lieferant: Apollo Scientific

OR310959-1G OR310959-500MG
APOSOR310959-1GEA 538 EUR
APOSOR310959-1G APOSOR310959-500MG
(4-Benzylphenyl)methanol
(4-Benzylphenyl)methanol
Formel: C₁₄H₁₄O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11846511
CAS-Nummer: 35714-20-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR