Methyl 2-hydroxy-5-isopropoxybenzoate

Lieferant: Apollo Scientific
Achtung

OR31012-1G OR31012-250MG
APOSOR31012-1GEA 111 EUR
APOSOR31012-1G APOSOR31012-250MG
Methyl 2-hydroxy-5-isopropoxybenzoate
Methyl 2-hydroxy-5-isopropoxybenzoate
Formel: C₁₁H₁₄O₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12546144
CAS-Nummer: 53434-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR