2,6-Dichloro-3-iodothiophene

Lieferant: Apollo Scientific

OR310093-1G OR310093-500MG OR310093-250MG
APOSOR310093-1GEA 260 EUR
APOSOR310093-1G APOSOR310093-500MG APOSOR310093-250MG
2,6-Dichloro-3-iodothiophene
2,6-Dichloro-3-iodothiophene
Formel: C₄HCl₂IS
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11844795
CAS-Nummer: 43225-59-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR