(2R)-1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR308064-1G
OR308064-5G
APOSOR308064-1GEA
64.4
EUR
APOSOR308064-1G
APOSOR308064-5G
(2R)-1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid 97%
(2R)-1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid
Formel:
C₁₀H₁₈N₂O₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02179106 CAS-Nummer: 278788-60-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...