Dimethyl bicyclo[2.2.2]octane-1,4-dicarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR307146-1G
OR307146-5G
APOSOR307146-1GEA
21.9
EUR
APOSOR307146-1G
APOSOR307146-5G
Dimethyl bicyclo[2.2.2]octane-1,4-dicarboxylate
Dimethyl bicyclo[2.2.2]octane-1,4-dicarboxylate
Formel:
C₁₂H₁₈O₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01312167 CAS-Nummer: 1459-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...