1H-Indole-3-boronic acid, pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole, 2-(1H-Indol-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR305200-1G OR305200-250MG
APOSOR305200-1GEA 184 EUR
APOSOR305200-1G APOSOR305200-250MG
1H-Indole-3-boronic acid, pinacol ester
1H-Indole-3-boronic acid, pinacol ester
Formel: C₁₄H₁₈BNO₂
Schmelzpunkt: 163…165 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09879158
CAS-Nummer: 937366-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR