4-Methoxy-1-benzothiophene-2-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 4-Methoxybenzo[b]thiophene-2-carboxylic acid

OR305045-250MG OR305045-1G
APOSOR305045-250MGEA 48 EUR
APOSOR305045-250MG APOSOR305045-1G
4-Methoxy-1-benzothiophene-2-carboxylic acid
4-Methoxy-1-benzothiophene-2-carboxylic acid
Formel: C₁₀H₈O₃S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07371550
CAS-Nummer: 476199-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR