Methyl potassium oxalate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium 2-methoxy-2-oxoacetate, Potassium methoxy(oxo)acetate, Potassium 2-methoxy-2-oxoethanoate

OR303344-1G OR303344-500MG
APOSOR303344-1GEA 23.2 EUR
APOSOR303344-1G APOSOR303344-500MG
Methyl potassium oxalate 98%
Methyl potassium oxalate
Formel: C₃H₃O₄·K
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20922789
CAS-Nummer: 10304-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR