3-{2-[(4-Chlorophenyl)thio]-5-nitrophenyl}acrylic acid
Lieferant: Apollo Scientific
OR30333-10G
OR30333-1G
OR30333-5G
APOSOR30333-10GEA
519
EUR
APOSOR30333-10G
APOSOR30333-1G
APOSOR30333-5G
3-{2-[(4-Chlorophenyl)thio]-5-nitrophenyl}acrylic acid
3-{2-[(4-Chlorophenyl)thio]-5-nitrophenyl}acrylic acid
Formel:
C₁₅H₁₀ClNO₄S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02090622 CAS-Nummer: 175278-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...