Allylamine, N-FMOC protected
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(9H-Fluoren-9-yl)methyl (prop-2-en-1-yl)carbamate, N-{[(9H-Fluoren-9-yl)methoxy]carbonyl}prop-2-en-1-amine
OR303191-1G
OR303191-500MG
APOSOR303191-1GEA
671
EUR
APOSOR303191-1G
APOSOR303191-500MG
Allylamine, N-FMOC protected
Allylamine, N-FMOC protected
Formel:
C₁₈H₁₇NO₂ Molekulargewicht: 279.333 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD14635842 CAS-Nummer: 856438-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...