2-(Piperidin-3-yl)thiazole 95%
Lieferant: Apollo Scientific
OR302415-1G
OR302415-250MG
APOSOR302415-1GEA
1350
EUR
APOSOR302415-1G
APOSOR302415-250MG
2-(Piperidin-3-yl)thiazole 95%
2-(Piperidin-3-yl)thiazole
Formel:
C₈H₁₂N₂S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09891523 CAS-Nummer: 630121-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...