2-Bromo-5-ethylpyridine

Lieferant: Apollo Scientific

OR302216-1G OR302216-250MG OR302216-5G
APOSOR302216-1GEA 271 EUR
APOSOR302216-1G APOSOR302216-250MG APOSOR302216-5G
2-Bromo-5-ethylpyridine
2-Bromo-5-ethylpyridine
Formel: C₇H₈BrN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11869552
CAS-Nummer: 19842-08-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR