Lithium trihydroxy(pyridin-2-yl)borate

Lieferant: Apollo Scientific

OR302190-5G OR302190-1G
APOSOR302190-5GEA 583 EUR
APOSOR302190-5G APOSOR302190-1G
Lithium trihydroxy(pyridin-2-yl)borate
Lithium trihydroxy(pyridin-2-yl)borate
Formel: C₅H₇BNO₃·Li
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16872059
CAS-Nummer: 1393822-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR