4-Cyano-2,6-dichlorobenzoic acid 95%

Lieferant: Apollo Scientific
Achtung

OR302095-1G OR302095-500MG
APOSOR302095-1GEA 870 EUR
APOSOR302095-1G APOSOR302095-500MG
4-Cyano-2,6-dichlorobenzoic acid 95%
4-Cyano-2,6-dichlorobenzoic acid
Formel: C₈H₃Cl₂NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD17926545
CAS-Nummer: 1258298-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR