Ethyl 4-bromobenzimidate hydrochloride, tech
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 4-bromobenzenecarboximidate hydrochloride, 4-Bromobenzimidic acid ethyl ester hydrochloride
OR300575-1G
OR300575-5G
APOSOR300575-1GEA
0
EUR
APOSOR300575-1G
APOSOR300575-5G
Ethyl 4-bromobenzimidate hydrochloride, tech
Ethyl 4-bromobenzimidate hydrochloride
Formel:
C₉H₁₀BrNO·ClH Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12131140 CAS-Nummer: 55368-83-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...