5-Bromo-3-cyclopropylpyrazin-2-amine

Lieferant: Apollo Scientific
Achtung

OR300237-1G OR300237-250MG
APOSOR300237-1GEA 719 EUR
APOSOR300237-1G APOSOR300237-250MG
5-Bromo-3-cyclopropylpyrazin-2-amine
5-Bromo-3-cyclopropylpyrazin-2-amine
Formel: C₇H₈BrN₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD31556420
CAS-Nummer: 1845754-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR