7-Methoxybenzo[b]thiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR2844-250MG
OR2844-1G
APOSOR2844-250MGEA
143
EUR
APOSOR2844-250MG
APOSOR2844-1G
7-Methoxybenzo[b]thiophene-2-carboxylic acid
7-Methoxybenzo[b]thiophene-2-carboxylic acid
Formel:
C₁₀H₈O₃S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01830203 CAS-Nummer: 88791-07-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...