{4-[(tert-Butoxycarbonyl)amino]tetrahydro-2H-pyran-4-yl}acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-4-(carboxymethyl)tetrahydro-2H-pyran, N-BOC protected, 4-[(tert-Butoxycarbonyl)amino]-4-(carboxymethyl)tetrahydro-2H-pyran

OR2668-100MG OR2668-250MG
APOSOR2668-100MGEA 87 EUR
APOSOR2668-100MG APOSOR2668-250MG
{4-[(tert-Butoxycarbonyl)amino]tetrahydro-2H-pyran-4-yl}acetic acid
{4-[(tert-Butoxycarbonyl)amino]tetrahydro-2H-pyran-4-yl}acetic acid
Formel: C₁₂H₂₁NO₅
Molekulargewicht: 259.299 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD09264196
CAS-Nummer: 946682-30-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR