1-(4-Chlorophenyl)-3-(dimethylamino)propan-1-one hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR25229-250MG
OR25229-1G
APOSOR25229-250MGEA
21.9
EUR
APOSOR25229-250MG
APOSOR25229-1G
1-(4-Chlorophenyl)-3-(dimethylamino)propan-1-one hydrochloride
1-(4-Chlorophenyl)-3-(dimethylamino)propan-1-one hydrochloride
Formel:
C₁₁H₁₄ClNO·ClH Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00084951 CAS-Nummer: 1798-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...