4-(Methylthio)-2-({[2-(methylthio)-3-pyridyl]carbonyl}amino)butanoic acid

Lieferant: Apollo Scientific

OR25155-1G OR25155-5G
APOSOR25155-1GEA 102 EUR
APOSOR25155-1G APOSOR25155-5G
4-(Methylthio)-2-({[2-(methylthio)-3-pyridyl]carbonyl}amino)butanoic acid
4-(Methylthio)-2-({[2-(methylthio)-3-pyridyl]carbonyl}amino)butanoic acid
Formel: C₁₂H₁₆N₂O₃S₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03407303
CAS-Nummer: 175201-70-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR