2,2'-Bithiophene-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR23231-1G OR23231-250MG
APOSOR23231-1GEA 184 EUR
APOSOR23231-1G APOSOR23231-250MG
2,2'-Bithiophene-5-carboxylic acid
2,2'-Bithiophene-5-carboxylic acid
Formel: C₉H₆O₂S₂
Schmelzpunkt: 177…178 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00159571
CAS-Nummer: 2060-55-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR