2-Amino-4-chloro-1,3-thiazole-5-carboxylic acid, 2-BOC protected
Lieferant: Apollo Scientific
|
Achtung
|
OR18817-1G
OR18817-250MG
APOSOR18817-1GEA
464
EUR
APOSOR18817-1G
APOSOR18817-250MG
2-Amino-4-chloro-1,3-thiazole-5-carboxylic acid, 2-BOC protected
2-Amino-4-chloro-1,3-thiazole-5-carboxylic acid, 2-BOC protected
Formel:
C₉H₁₁ClN₂O₄S Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD24476508 CAS-Nummer: 612541-07-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...