4'-(Hydroxymethyl)acetophenone
Lieferant: Apollo Scientific
Synonyme:
1-(4-(Hydroxymethyl)phenyl)ethanone, 1-[4-(Hydroxymethyl)phenyl]ethan-1-one, 4-Acetylbenzyl alcohol
OR18571-1G
OR18571-250MG
APOSOR18571-1GEA
238
EUR
APOSOR18571-1G
APOSOR18571-250MG
4'-(Hydroxymethyl)acetophenone
4'-(Hydroxymethyl)acetophenone
Formel:
C₉H₁₀O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06202822 CAS-Nummer: 75633-63-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...