3-Amino-5-nitrobenzo[b]thiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR183423-1G
OR183423-250MG
APOSOR183423-1GEA
364
EUR
APOSOR183423-1G
APOSOR183423-250MG
3-Amino-5-nitrobenzo[b]thiophene-2-carboxylic acid
3-Amino-5-nitrobenzo[b]thiophene-2-carboxylic acid
Formel:
C₉H₆N₂O₄S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD20924150 CAS-Nummer: 160041-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...